Assay Details
Gene Symbol: ctss2.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017661.1 | NP_001017661.1 | 2 - 3 | 158 | 82 | |
XM_009292324.3 | XP_009290599.1 | 1 - 2 | 156 | 82 | ||
GenBank mRNA | BC093339.1 | - | 2 - 3 | 158 | 82 | |
BC096862.1 | - | 2 - 3 | 158 | 82 | ||
BC164362.1 | - | 1 - 2 | 154 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
