Assay Details
Gene Symbol: ctss2.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017661.1 | NP_001017661.1 | 6 - 7 | 827 | 65 | |
XM_009292324.3 | XP_009290599.1 | 5 - 6 | 825 | 65 | ||
GenBank mRNA | BC093339.1 | - | 6 - 7 | 827 | 65 | |
BC096862.1 | - | 6 - 7 | 827 | 65 | ||
BC164362.1 | - | 5 - 6 | 823 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
