Assay Details
| Gene Symbol: prkar1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017732.1 | NP_001017732.1 | 8 - 9 | 879 | 64 | |
| GenBank mRNA | BC093222.1 | - | 8 - 9 | 879 | 64 | |
More Information
Set Membership: |
|

prkar1ab
Genomic Map