Assay Details
| Gene Symbol: prkar1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017732.1 | NP_001017732.1 | 10 - 11 | 1088 | 60 | |
| GenBank mRNA | BC093222.1 | - | 10 - 11 | 1088 | 60 | |
More Information
Set Membership: |
|

prkar1ab
Genomic Map