Assay Details
| Gene Symbol: pdlim3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024376.1 | NP_001019547.1 | 2 - 3 | 282 | 152 | |
| GenBank mRNA | BC095249.1 | - | 2 - 3 | 282 | 152 | |
More Information
Set Membership: |
|

pdlim3a
Genomic Map