Assay Details
Gene Symbol: pdlim3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024376.1 | NP_001019547.1 | 4 - 5 | 539 | 88 | |
GenBank mRNA | BC095249.1 | - | 4 - 5 | 539 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
