Assay Details
Gene Symbol: slc6a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039972.1 | NP_001035061.1 | 3 - 4 | 827 | 78 | |
XM_005157654.4 | XP_005157711.1 | 6 - 7 | 2005 | 78 | ||
XM_005157655.4 | XP_005157712.1 | 6 - 7 | 1928 | 78 | ||
XM_009291668.3 | XP_009289943.1 | 7 - 8 | 2197 | 78 | ||
XM_009291669.3 | XP_009289944.1 | 7 - 8 | 2115 | 78 | ||
XM_009291670.3 | XP_009289945.1 | 5 - 6 | 1736 | 78 | ||
XM_009291671.3 | XP_009289946.1 | 7 - 8 | 1493 | 78 | ||
XM_009291672.3 | XP_009289947.1 | 5 - 6 | 1654 | 78 | ||
GenBank mRNA | DQ285098.1 | - | 3 - 4 | 827 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
