Assay Details
Gene Symbol: slc6a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039972.1 | NP_001035061.1 | 4 - 5 | 959 | 84 | |
XM_005157654.4 | XP_005157711.1 | 7 - 8 | 2137 | 84 | ||
XM_005157655.4 | XP_005157712.1 | 7 - 8 | 2060 | 84 | ||
XM_009291668.3 | XP_009289943.1 | 8 - 9 | 2329 | 84 | ||
XM_009291669.3 | XP_009289944.1 | 8 - 9 | 2247 | 84 | ||
XM_009291670.3 | XP_009289945.1 | 6 - 7 | 1868 | 84 | ||
XM_009291671.3 | XP_009289946.1 | 8 - 9 | 1625 | 84 | ||
XM_009291672.3 | XP_009289947.1 | 6 - 7 | 1786 | 84 | ||
XM_009291673.3 | XP_009289948.1 | 4 - 5 | 708 | 84 | ||
GenBank mRNA | BC163766.1 | - | 4 - 5 | 959 | 84 | |
BC163777.1 | - | 4 - 5 | 959 | 84 | ||
DQ285098.1 | - | 4 - 5 | 959 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
