Assay Details
| Gene Symbol: slc6a4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039972.1 | NP_001035061.1 | 7 - 8 | 1335 | 75 | |
| XM_005157654.4 | XP_005157711.1 | 10 - 11 | 2513 | 75 | ||
| XM_005157655.4 | XP_005157712.1 | 10 - 11 | 2436 | 75 | ||
| XM_009291668.3 | XP_009289943.1 | 11 - 12 | 2705 | 75 | ||
| XM_009291669.3 | XP_009289944.1 | 11 - 12 | 2623 | 75 | ||
| XM_009291670.3 | XP_009289945.1 | 9 - 10 | 2244 | 75 | ||
| XM_009291671.3 | XP_009289946.1 | 11 - 12 | 2001 | 75 | ||
| XM_009291672.3 | XP_009289947.1 | 9 - 10 | 2162 | 75 | ||
| XM_009291673.3 | XP_009289948.1 | 7 - 8 | 1084 | 75 | ||
| GenBank mRNA | BC163766.1 | - | 7 - 8 | 1335 | 75 | |
| BC163777.1 | - | 7 - 8 | 1335 | 75 | ||
| DQ285098.1 | - | 7 - 8 | 1335 | 75 | ||
More Information
Set Membership: |
|

slc6a4a
Genomic Map