Assay Details
Gene Symbol: slc6a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039972.1 | NP_001035061.1 | 9 - 10 | 1585 | 68 | |
XM_005157654.4 | XP_005157711.1 | 12 - 13 | 2763 | 68 | ||
XM_005157655.4 | XP_005157712.1 | 12 - 13 | 2686 | 68 | ||
XM_009291668.3 | XP_009289943.1 | 13 - 14 | 2955 | 68 | ||
XM_009291669.3 | XP_009289944.1 | 13 - 14 | 2873 | 68 | ||
XM_009291670.3 | XP_009289945.1 | 11 - 12 | 2494 | 68 | ||
XM_009291671.3 | XP_009289946.1 | 13 - 14 | 2251 | 68 | ||
XM_009291672.3 | XP_009289947.1 | 11 - 12 | 2412 | 68 | ||
XM_009291673.3 | XP_009289948.1 | 9 - 10 | 1334 | 68 | ||
GenBank mRNA | BC163766.1 | - | 9 - 10 | 1585 | 68 | |
BC163777.1 | - | 9 - 10 | 1585 | 68 | ||
DQ285098.1 | - | 9 - 10 | 1585 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
