Assay Details
| Gene Symbol: ppp1r13bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044824.2 | NP_001038289.2 | 14 - 15 | 2828 | 105 | |
| XM_005160501.4 | XP_005160558.1 | 13 - 14 | 2830 | 105 | ||
| XM_009294640.3 | XP_009292915.1 | 14 - 15 | 2981 | 105 | ||
| XM_009294641.3 | XP_009292916.1 | 14 - 15 | 2958 | 105 | ||
| XM_009294642.3 | XP_009292917.1 | 15 - 16 | 3231 | 105 | ||
| XM_009294643.3 | XP_009292918.1 | 12 - 13 | 2649 | 105 | ||
| GenBank mRNA | BC139881.1 | - | 14 - 15 | 2828 | 105 | |
More Information
Set Membership: |
|

ppp1r13bb
Genomic Map