Assay Details
| Gene Symbol: ppp1r13bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044824.2 | NP_001038289.2 | 6 - 7 | 607 | 68 | |
| XM_005160501.4 | XP_005160558.1 | 5 - 6 | 609 | 68 | ||
| XM_009294640.3 | XP_009292915.1 | 5 - 6 | 610 | 68 | ||
| XM_009294641.3 | XP_009292916.1 | 5 - 6 | 611 | 68 | ||
| XM_009294642.3 | XP_009292917.1 | 6 - 7 | 860 | 68 | ||
| XM_009294643.3 | XP_009292918.1 | 3 - 4 | 278 | 68 | ||
| GenBank mRNA | BC139881.1 | - | 6 - 7 | 607 | 68 | |
More Information
Set Membership: |
|

ppp1r13bb
Genomic Map