Assay Details
Gene Symbol: neu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044909.2 | NP_001038374.1 | 3 - 4 | 597 | 58 | |
XM_009294220.3 | XP_009292495.1 | 3 - 4 | 542 | 58 | ||
GenBank mRNA | BC133832.1 | - | 3 - 4 | 597 | 58 | |
LC373154.1 | - | 3 - 4 | 529 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
