Assay Details
Gene Symbol: neu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044909.2 | NP_001038374.1 | 5 - 6 | 1000 | 105 | |
XM_009294220.3 | XP_009292495.1 | 5 - 6 | 945 | 105 | ||
GenBank mRNA | BC133832.1 | - | 5 - 6 | 1000 | 105 | |
LC373154.1 | - | 5 - 6 | 932 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
