Assay Details
| Gene Symbol: shdb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045077.2 | NP_001038542.1 | 4 - 5 | 867 | 107 | |
| XM_005170476.4 | XP_005170533.1 | 5 - 6 | 1314 | 107 | ||
| XM_009296270.3 | XP_009294545.1 | 6 - 7 | 1393 | 107 | ||
| XM_021469342.1 | XP_021325017.1 | 6 - 7 | 1450 | 107 | ||
| GenBank mRNA | BC124504.1 | - | 5 - 6 | 1288 | 107 | |
More Information
Set Membership: |
|

shdb
Genomic Map