Assay Details
| Gene Symbol: znf384b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045163.1 | NP_001038628.1 | 5 - 6 | 619 | 65 | |
| XM_009292249.3 | XP_009290524.1 | 5 - 6 | 1241 | 65 | ||
| XM_009292250.3 | XP_009290525.1 | 5 - 6 | 1059 | 65 | ||
| XM_009292251.3 | XP_009290526.1 | 5 - 6 | 923 | 65 | ||
| GenBank mRNA | BC129180.1 | - | 5 - 6 | 621 | 65 | |
More Information
Set Membership: |
|

znf384b
Genomic Map