Assay Details
| Gene Symbol: wnt9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045363.1 | NP_001038828.1 | 3 - 4 | 293 | 122 | |
| XM_005163661.4 | XP_005163718.1 | 2 - 3 | 402 | 230 | ||
| GenBank mRNA | BC122352.1 | - | 3 - 4 | 293 | 122 | |
| BC164356.1 | - | 3 - 4 | 277 | 122 | ||
| FJ231750.1 | - | 2 - 3 | 410 | 230 | ||
| GQ927508.1 | - | 3 - 4 | 293 | 122 | ||
More Information
Set Membership: |
|

wnt9a
Genomic Map