Assay Details
| Gene Symbol: rragd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099424.2 | NP_001092894.2 | 1 - 2 | 361 | 80 | |
| XM_021467215.1 | XP_021322890.1 | 1 - 2 | 387 | 80 | ||
| GenBank mRNA | BC142594.1 | - | 1 - 2 | 361 | 80 | |
More Information
Set Membership: |
|

rragd
Genomic Map