Assay Details
| Gene Symbol: ube2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103116.1 | NP_001096586.1 | 2 - 3 | 409 | 73 | |
| XM_005159506.4 | XP_005159563.1 | 2 - 3 | 262 | 73 | ||
| GenBank mRNA | BC151907.1 | - | 2 - 3 | 409 | 73 | |
More Information
Set Membership: |
|

ube2e1
Genomic Map