Assay Details
| Gene Symbol: tent5ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103125.1 | NP_001096595.1 | 2 - 3 | 823 | 95 | |
| GenBank mRNA | BC151974.1 | - | 2 - 3 | 823 | 95 | |
More Information
Set Membership: |
|

tent5ab
Genomic Map