Assay Details
| Gene Symbol: senp6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021468556.1 | XP_021324231.1 | 15 - 16 | 2311 | 56 | |
| XM_021468557.1 | XP_021324232.1 | 14 - 15 | 2247 | 56 | ||
| XM_021468558.1 | XP_021324233.1 | 15 - 16 | 2298 | 56 | ||
| XM_021468559.1 | XP_021324234.1 | 15 - 16 | 2308 | 56 | ||
| XM_021468560.1 | XP_021324235.1 | 14 - 15 | 2217 | 56 | ||
| XM_021468561.1 | XP_021324236.1 | 14 - 15 | 2204 | 56 | ||
| GenBank mRNA | BC154420.1 | - | 2 - 3 | 320 | 56 | |
More Information
Set Membership: |
|

senp6a
Genomic Map