Assay Details
| Gene Symbol: senp6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021468556.1 | XP_021324231.1 | 21 - 22 | 2999 | 67 | |
| XM_021468557.1 | XP_021324232.1 | 20 - 21 | 2935 | 67 | ||
| XM_021468558.1 | XP_021324233.1 | 21 - 22 | 2986 | 67 | ||
| XM_021468559.1 | XP_021324234.1 | 21 - 22 | 2996 | 67 | ||
| XM_021468560.1 | XP_021324235.1 | 20 - 21 | 2905 | 67 | ||
| XM_021468561.1 | XP_021324236.1 | 20 - 21 | 2892 | 67 | ||
| GenBank mRNA | BC134226.1 | - | 5 - 6 | 463 | 67 | |
| BC154420.1 | - | 8 - 9 | 1008 | 67 | ||
More Information
Set Membership: |
|

senp6a
Genomic Map