Assay Details
| Gene Symbol: tdgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328408.1 | NP_001315337.1 | 5 - 6 | 807 | 96 | |
| NM_131092.2 | NP_571167.2 | 6 - 7 | 828 | 117 | ||
| GenBank mRNA | AF041440.1 | - | 6 - 7 | 558 | 117 | |
| AF041441.1 | - | 5 - 6 | 537 | 96 | ||
| BC095007.1 | - | 5 - 6 | 803 | 96 | ||
| BC152111.1 | - | 5 - 6 | 492 | 96 | ||
| BC152235.1 | - | 5 - 6 | 546 | 96 | ||
| BC165306.1 | - | 5 - 6 | 487 | 96 | ||
More Information
Set Membership: |
|

tdgf1
Genomic Map