Assay Details
Gene Symbol: opn1sw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131319.1 | NP_571394.1 | 4 - 5 | 978 | 124 | |
GenBank mRNA | AF104903.1 | - | 2 - 3 | 420 | 124 | |
AF109373.1 | - | 4 - 5 | 978 | 124 | ||
BC060894.1 | - | 4 - 5 | 964 | 124 | ||
BC067683.1 | - | 4 - 5 | 959 | 124 | ||
BC153466.1 | - | 4 - 5 | 977 | 124 | ||
KT008399.1 | - | 4 - 5 | 917 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
