Assay Details
| Gene Symbol: guca1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131871.1 | NP_571946.1 | 2 - 3 | 582 | 73 | |
| GenBank mRNA | AY044456.1 | - | 2 - 3 | 579 | 73 | |
| AY050502.1 | - | 2 - 3 | 582 | 73 | ||
| BC124761.1 | - | 2 - 3 | 578 | 73 | ||
More Information
Set Membership: |
|

guca1b
Genomic Map