Assay Details
| Gene Symbol: esr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174862.3 | NP_777287.2 | 1 - 2 | 448 | 81 | |
| XM_005156971.4 | XP_005157028.1 | 2 - 3 | 757 | 81 | ||
| XM_005156972.4 | XP_005157029.1 | 3 - 4 | 931 | 81 | ||
| XM_005156973.4 | XP_005157030.1 | 2 - 3 | 468 | 81 | ||
| GenBank mRNA | AF349413.3 | - | 2 - 3 | 467 | 81 | |
| AJ275911.3 | - | 2 - 3 | 488 | 81 | ||
| AJ414566.1 | - | 2 - 3 | 468 | 81 | ||
| BC086848.1 | - | 1 - 2 | 448 | 81 | ||
| BC164792.1 | - | 1 - 2 | 423 | 81 | ||
More Information
Set Membership: |
|

esr2b
Genomic Map