Assay Details
| Gene Symbol: cd63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199543.1 | NP_955837.1 | 7 - 8 | 725 | 58 | |
| GenBank mRNA | BC056698.1 | - | 7 - 8 | 725 | 58 | |
| BC064300.1 | - | 7 - 8 | 763 | 58 | ||
More Information
Set Membership: |
|

cd63
Genomic Map