Assay Details
| Gene Symbol: lgi1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199627.2 | NP_955921.2 | 3 - 4 | 475 | 97 | |
| GenBank mRNA | BC056753.1 | - | 3 - 4 | 458 | 97 | |
| BC065665.1 | - | 3 - 4 | 439 | 97 | ||
| CU638810.1 | - | 3 - 4 | 320 | 97 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  Mammalian Gene Collection  Detect Genomic DNA  Danio rerio  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 lgi1a
lgi1a Genomic Map
Genomic Map Back To Top
 Back To Top