Assay Details
| Gene Symbol: lgi1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199627.2 | NP_955921.2 | 7 - 8 | 969 | 74 | |
| GenBank mRNA | AY309261.1 | - | 7 - 8 | 911 | 74 | |
| BC056753.1 | - | 7 - 8 | 952 | 74 | ||
| BC065665.1 | - | 7 - 8 | 933 | 74 | ||
| CU468780.1 | - | 7 - 8 | 801 | 74 | ||
More Information
Set Membership: |
|

lgi1a
Genomic Map