Assay Details
| Gene Symbol: cers5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199628.1 | NP_955922.1 | 2 - 3 | 358 | 62 | |
| XM_021469303.1 | XP_021324978.1 | 2 - 3 | 766 | 62 | ||
| GenBank mRNA | BC053143.1 | - | 2 - 3 | 358 | 62 | |
More Information
Set Membership: |
|

cers5
Genomic Map