Assay Details
| Gene Symbol: rab5if | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199850.2 | NP_956144.2 | 2 - 3 | 367 | 82 | |
| GenBank mRNA | BC042320.1 | - | 2 - 3 | 347 | 82 | |
| BC164934.1 | - | 2 - 3 | 248 | 82 | ||
| CT613988.2 | - | 2 - 3 | 333 | 82 | ||
| EH540625.1 | - | 2 - 3 | 421 | 82 | ||
More Information
Set Membership: |
|

rab5if
Genomic Map