Assay Details
| Gene Symbol: rab5if | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199850.2 | NP_956144.2 | 3 - 4 | 493 | 72 | |
| GenBank mRNA | BC042320.1 | - | 3 - 4 | 473 | 72 | |
| BC164934.1 | - | 3 - 4 | 374 | 72 | ||
| CT613988.2 | - | 3 - 4 | 459 | 72 | ||
| EH540625.1 | - | 3 - 4 | 547 | 72 | ||
More Information
Set Membership: |
|

rab5if
Genomic Map