Assay Details
Gene Symbol: atp5f1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199968.1 | NP_956262.1 | 2 - 3 | 331 | 85 | |
GenBank mRNA | BC049434.1 | - | 2 - 3 | 375 | 85 | |
BC059627.1 | - | 2 - 3 | 331 | 85 | ||
BC065042.1 | - | 2 - 3 | 325 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
