Assay Details
| Gene Symbol: rps3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200059.2 | NP_956353.1 | 1 - 2 | 88 | 88 | |
| GenBank mRNA | AY648734.1 | - | 1 - 2 | 69 | 88 | |
| BC059543.1 | - | 1 - 2 | 67 | 88 | ||
| BC078649.1 | - | 1 - 2 | 88 | 88 | ||
| CK357107.1 | - | 1 - 2 | 102 | 88 | ||
More Information
Set Membership: |
|

rps3a
Genomic Map