Assay Details
| Gene Symbol: calm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200082.1 | NP_956376.1 | 4 - 5 | 345 | 60 | |
| GenBank mRNA | BC059500.1 | - | 4 - 5 | 345 | 60 | |
| BC065426.1 | - | 4 - 5 | 339 | 60 | ||
More Information
Set Membership: |
|

calm1b
Genomic Map