Assay Details
| Gene Symbol: dmac2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200218.1 | NP_956512.1 | 2 - 3 | 165 | 120 | |
| GenBank mRNA | BC046029.1 | - | 2 - 3 | 165 | 120 | |
More Information
Set Membership: |
|

dmac2l
Genomic Map