Assay Details
| Gene Symbol: slc35d1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200475.2 | NP_956769.2 | 5 - 6 | 537 | 109 | |
| GenBank mRNA | BC055186.1 | - | 5 - 6 | 537 | 109 | |
More Information
Set Membership: |
|

slc35d1b
Genomic Map