Assay Details
| Gene Symbol: arpc5la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200578.2 | NP_956872.1 | 3 - 4 | 583 | 88 | |
| GenBank mRNA | BC056697.1 | - | 3 - 4 | 599 | 88 | |
| BC065664.1 | - | 3 - 4 | 583 | 88 | ||
More Information
Set Membership: |
|

arpc5la
Genomic Map