Assay Details
Gene Symbol: med30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200717.1 | NP_957011.1 | 4 - 5 | 510 | 91 | |
XM_009294453.3 | XP_009292728.1 | 3 - 4 | 435 | 91 | ||
GenBank mRNA | BC059478.1 | - | 4 - 5 | 510 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
