Assay Details
| Gene Symbol: gnai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200971.1 | NP_957265.1 | 8 - 9 | 1343 | 69 | |
| GenBank mRNA | BC053164.1 | - | 8 - 9 | 1343 | 69 | |
| BC154335.1 | - | 8 - 9 | 1202 | 69 | ||
More Information
Set Membership: |
|

gnai1
Genomic Map