Assay Details
| Gene Symbol: gps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200973.3 | NP_957267.1 | 11 - 12 | 1551 | 60 | |
| XM_005165131.3 | XP_005165188.1 | 11 - 12 | 1172 | 60 | ||
| XM_017356047.2 | XP_017211536.1 | 11 - 12 | 1413 | 60 | ||
| XM_021476040.1 | XP_021331715.1 | 12 - 13 | 1185 | 60 | ||
| GenBank mRNA | BC054126.1 | - | 11 - 12 | 1136 | 60 | |
| BC096776.1 | - | 11 - 12 | 1552 | 60 | ||
| BC164184.1 | - | 10 - 11 | 1028 | 60 | ||
More Information
Set Membership: |
|

gps2
Genomic Map