Assay Details
Gene Symbol: rpl34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201122.1 | NP_957416.1 | 2 - 3 | 89 | 110 | |
XM_005166535.3 | XP_005166592.1 | 2 - 3 | 310 | 110 | ||
GenBank mRNA | BC049023.1 | - | 2 - 3 | 89 | 110 | |
BC062280.1 | - | 1 - 2 | 73 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
