Assay Details
| Gene Symbol: hibadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201160.1 | NP_957454.1 | 2 - 3 | 286 | 68 | |
| GenBank mRNA | BC055573.1 | - | 2 - 3 | 286 | 68 | |
More Information
Set Membership: |
|

hibadhb
Genomic Map