Assay Details
| Gene Symbol: ywhaqb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201484.1 | NP_958892.1 | 5 - 6 | 841 | 68 | |
| GenBank mRNA | AB194124.1 | - | 5 - 6 | 814 | 68 | |
| BC044412.1 | - | 5 - 6 | 841 | 68 | ||
More Information
Set Membership: |
|

ywhaqb
Genomic Map