Assay Details
| Gene Symbol: arpc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201509.1 | NP_958917.1 | 2 - 3 | 359 | 89 | |
| GenBank mRNA | AY394953.1 | - | 2 - 3 | 358 | 89 | |
| BC049407.1 | - | 2 - 3 | 359 | 89 | ||
More Information
Set Membership: |
|

arpc5b
Genomic Map