Assay Details
Gene Symbol: drap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205541.2 | NP_991104.2 | 2 - 3 | 307 | 60 | |
GenBank mRNA | BC065342.1 | - | 2 - 3 | 240 | 60 | |
BC067379.1 | - | 2 - 3 | 234 | 60 | ||
EH455821.1 | - | 2 - 3 | 274 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
