Assay Details
Gene Symbol: arl8ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205566.1 | NP_991129.1 | 1 - 2 | 175 | 146 | |
GenBank mRNA | BC065942.1 | - | 1 - 2 | 175 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
