Assay Details
Gene Symbol: mknk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212723.1 | NP_997888.1 | 6 - 7 | 632 | 120 | |
XM_021479017.1 | XP_021334692.1 | 6 - 7 | 649 | 120 | ||
GenBank mRNA | BC045391.1 | - | 6 - 7 | 632 | 120 | |
BC165349.1 | - | 5 - 6 | 450 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
