Assay Details
| Gene Symbol: rpl36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212952.2 | NP_998117.1 | 2 - 3 | 130 | 69 | |
| XM_005170351.4 | XP_005170408.1 | 1 - 2 | 99 | 69 | ||
| GenBank mRNA | AY561518.1 | - | 1 - 2 | 99 | 69 | |
| BC071384.1 | - | 2 - 3 | 130 | 69 | ||
More Information
Set Membership: |
|

rpl36
Genomic Map