Assay Details
| Gene Symbol: zfand5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213039.1 | NP_998204.1 | 2 - 3 | 288 | 78 | |
| XM_021476057.1 | XP_021331732.1 | 2 - 3 | 470 | 78 | ||
| GenBank mRNA | BC059673.1 | - | 2 - 3 | 288 | 78 | |
More Information
Set Membership: |
|

zfand5a
Genomic Map